Search
Search

CHI3L1| Gene Symbol: CHI3L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276.2 | NP_001267.2 | 5, 6 | 642 | ||
| GenBank mRNA | AK130142.1 | - | 5, 6 | 552 | ||
| AK130200.1 | - | 5, 6 | 552 | |||
| AK225266.1 | - | 5, 6 | 551 | |||
| AK225584.1 | - | 5, 6 | 551 | |||
| AK312911.1 | - | 5, 6 | 625 | |||
| BC008568.1 | - | 5, 6 | 588 | 4212748 | ||
| BC038354.1 | - | 5, 6 | 529 | 4212419 | ||
| BC039132.1 | - | 5, 6 | 538 | 4183798 | ||
| BT007223.1 | - | 5, 6 | 471 | |||
| M80927.1 | - | 5, 6 | 542 | |||