Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033133.4 | NP_149124.3 | 4 | 1394 | ||
XM_006721701.3 | XP_006721764.1 | 4 | 1418 | |||
XM_011524340.2 | XP_011522642.1 | 4 | 1379 | |||
GenBank mRNA | AK026876.1 | - | 4 | 1351 | ||
AK097833.1 | - | 4 | 1242 | |||
AK098384.1 | - | 4 | 1340 | |||
AK294179.1 | - | 3 | 667 | |||
AK295351.1 | - | 5 | 975 | |||
AK308696.1 | - | 2 | 480 | |||
BC001362.2 | - | 4 | 1331 | 3051112 | ||
BC006392.2 | - | 4 | 1353 | 4126696 | ||
BC011046.1 | - | 4 | 1315 | 4179083 | ||
BC028040.1 | - | 4 | 1355 | 5248370 | ||
M19650.1 | - | 3 | 1203 |