Search
Search

CRABP2| Gene Symbol: CRABP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199723.1 | NP_001186652.1 | 3 | 395 | ||
| NM_001878.3 | NP_001869.1 | 2 | 408 | |||
| XM_017000343.1 | XP_016855832.1 | 5 | 5822 | |||
| GenBank mRNA | AB593017.1 | - | 2 | 324 | ||
| AK312007.1 | - | 2 | 324 | |||
| BC001109.2 | - | 2 | 310 | 2988143 | ||
| BP231157.1 | - | 2 | 408 | |||
| BT019827.1 | - | 2 | 187 | |||
| CR450357.1 | - | 2 | 187 | |||
| M68867.1 | - | 2 | 285 | |||