Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NKX2-5 SIRNA Assays ›
          Gene Symbol:
          NKX2-5
          Gene Name:
          NK2 homeobox 5
          Gene Aliases:
          CHNG5, CSX, CSX1, HLHS2, NKX2.5, NKX2E, NKX4-1, VSD3
          Chromosome Location:
          Chr.5: 173232104 - 173235312 on Build GRCh38
          UniGene ID:
          Hs.54473
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS102432
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NKX2-5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166175.1 NP_001159647.1 1 543
          NM_001166176.1 NP_001159648.1 1 543
          NM_004387.3 NP_004378.1 1 543
          XM_017009071.1 XP_016864560.1 1 437
          GenBank mRNA AB021133.1 - 1 314
          AK290615.1 - 1 435
          AK297844.1 - 1 437
          AK307218.1 - 1 437
          AK307247.1 - 1 495
          AK307249.1 - 1 436
          AK309495.1 - 1 437
          BC025711.1 - 1 421 5225103
          DA559427.1 - 1 543
          KU178001.1 - 1 314
          KU178002.1 - 1 314
          U34962.1 - 1 490

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          vasculogenesis
          heart looping
          heart morphogenesis
          outflow tract septum morphogenesis
          cardiac conduction system development
          bundle of His development
          Purkinje myocyte differentiation
          cardiac ventricle formation
          right ventricular cardiac muscle tissue morphogenesis
          ventricular trabecula myocardium morphogenesis
          apoptotic process involved in heart morphogenesis
          septum secundum development
          proepicardium development
          pulmonary myocardium development
          transcription from RNA polymerase II promoter
          adult heart development
          positive regulation of cell proliferation
          negative regulation of cardiac muscle cell apoptotic process
          positive regulation of transcription via serum response element binding
          positive regulation of sodium ion transport
          negative regulation of myotube differentiation
          hemopoiesis
          cell differentiation
          BMP signaling pathway
          thyroid gland development
          embryonic heart tube development
          negative regulation of apoptotic process
          sarcomere organization
          positive regulation of neuron differentiation
          positive regulation of heart contraction
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          spleen development
          positive regulation of cardioblast differentiation
          ventricular cardiac myofibril assembly
          cardiac muscle cell differentiation
          cardiac muscle tissue morphogenesis
          atrial cardiac muscle cell development
          ventricular cardiac muscle cell development
          regulation of cardiac muscle contraction
          pharyngeal system development
          cardiac muscle cell proliferation
          regulation of cardiac muscle cell proliferation
          cardiac muscle contraction
          canonical Wnt signaling pathway
          positive regulation of transcription initiation from RNA polymerase II promoter
          heart trabecula formation
          ventricular septum morphogenesis
          atrial septum morphogenesis
          atrioventricular node cell development
          atrioventricular node cell fate commitment
          embryonic heart tube left/right pattern formation
          negative regulation of canonical Wnt signaling pathway
          regulation of cardiac conduction
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription cofactor activity
          transcriptional activator activity, RNA polymerase II transcription factor binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          protein binding
          transcription factor binding
          serum response element binding
          protein homodimerization activity
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0