Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CTNNA1 SIRNA Assays ›
          Gene Symbol:
          CTNNA1
          Gene Name:
          catenin alpha 1
          Gene Aliases:
          CAP102, MDPT2
          Chromosome Location:
          Chr.5: 138753386 - 138935034 on Build GRCh38
          UniGene ID:
          Hs.445981
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS102452
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price 459.00
          Your Price
          Online offer:
          459.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CTNNA1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001290307.2 NP_001277236.1 12, 13 1873
          NM_001290309.2 NP_001277238.1 11, 12 1677
          NM_001290310.2 NP_001277239.1 12, 13 1710
          NM_001290312.1 NP_001277241.1 6, 7 1193
          NM_001323982.1 NP_001310911.1 13, 14 2343
          NM_001323983.1 NP_001310912.1 12, 13 2025
          NM_001323984.1 NP_001310913.1 13, 14 1891
          NM_001323985.1 NP_001310914.1 12, 13 1873
          NM_001323986.1 NP_001310915.1 11, 12 1780
          NM_001323987.1 NP_001310916.1 6, 7 882
          NM_001323988.1 NP_001310917.1 9, 10 1130
          NM_001323989.1 NP_001310918.1 7, 8 978
          NM_001323990.1 NP_001310919.1 8, 9 1342
          NM_001323991.1 NP_001310920.1 7, 8 1289
          NM_001323992.1 NP_001310921.1 9, 10 1122
          NM_001323993.1 NP_001310922.1 9, 10 1436
          NM_001323994.1 NP_001310923.1 6, 7 891
          NM_001323995.1 NP_001310924.1 9, 10 1106
          NM_001323996.1 NP_001310925.1 9, 10 1441
          NM_001323997.1 NP_001310926.1 9, 10 1125
          NM_001323998.1 NP_001310927.1 9, 10 1417
          NM_001323999.1 NP_001310928.1 9, 10 1414
          NM_001324000.1 NP_001310929.1 7, 8 1286
          NM_001324001.1 NP_001310930.1 6, 7 1193
          NM_001324002.1 NP_001310931.1 6, 7 1193
          NM_001324003.1 NP_001310932.1 8, 9 1342
          NM_001324004.1 NP_001310933.1 6, 7 882
          NM_001324005.1 NP_001310934.1 9, 10 1417
          NM_001324006.1 NP_001310935.1 10, 11 1164
          NM_001324007.1 NP_001310936.1 9, 10 1400
          NM_001324008.1 NP_001310937.1 7, 8 940
          NM_001324009.1 NP_001310938.1 7, 8 1251
          NM_001324010.1 NP_001310939.1 3, 4 555
          NM_001324011.1 NP_001310940.1 8, 9 1013
          NM_001324012.1 NP_001310941.1 4, 5 648
          NM_001324013.1 NP_001310942.1 7, 8 1222
          NM_001903.4 NP_001894.2 12, 13 1873
          GenBank mRNA AK094151.1 - 9, 10 1082
          AK296708.1 - 12, 13 1681
          AK300439.1 - 11, 12 1644
          AK304267.1 - 12, 13 1768
          BC000385.2 - 12, 13 1811 2821111
          BC031262.1 - 6, 7 772 5274351
          D13866.1 - 12, 13 1812
          D14705.1 - 12, 13 1837
          HQ589335.1 - 6, 7 1027
          L23805.1 - 11, 12 1753
          U03100.1 - 11, 12 1753

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          actin filament organization
          cell adhesion
          establishment or maintenance of cell polarity
          negative regulation of neuroblast proliferation
          aging
          male gonad development
          gap junction assembly
          axon regeneration
          adherens junction organization
          cellular protein localization
          odontogenesis of dentin-containing tooth
          apical junction assembly
          response to estrogen
          positive regulation of smoothened signaling pathway
          positive regulation of muscle cell differentiation
          protein heterooligomerization
          cellular response to indole-3-methanol
          epithelial cell-cell adhesion
          negative regulation of cell motility
          negative regulation of integrin-mediated signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          structural molecule activity
          protein binding
          beta-catenin binding
          vinculin binding
          poly(A) RNA binding
          gamma-catenin binding
          cadherin binding
          actin filament binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-4p4qf:80/100.66.72.111:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0