Search
Search

DAD1| Gene Symbol: DAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001344.3 | NP_001335.1 | 1 | 121 | ||
| GenBank mRNA | AK223129.1 | - | 1 | 108 | ||
| BC007403.2 | - | 1 | 93 | 2988797 | ||
| BC009798.1 | - | 1 | 95 | 4284779 | ||
| BP198316.1 | - | 1 | 121 | |||
| CR407682.1 | - | 1 | 41 | |||
| CR542204.1 | - | 1 | 41 | |||
| D15057.1 | - | 1 | 107 | |||