Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DLG3 SIRNA Assays ›
          Gene Symbol:
          DLG3
          Gene Name:
          discs large MAGUK scaffold protein 3
          Gene Aliases:
          MRX, MRX90, NEDLG, PPP1R82, SAP102, XLMR
          Chromosome Location:
          Chr.X: 70444850 - 70505490 on Build GRCh38
          UniGene ID:
          Hs.721586
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS102796
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DLG3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001166278.1 NP_001159750.1 7, 8 726
          NM_020730.2 NP_065781.1 9, 10 1269
          NM_021120.3 NP_066943.2 14, 15 2214
          XM_005262248.3 XP_005262305.1 9, 10 866
          XM_006724625.2 XP_006724688.1 15, 16 2315
          XM_006724626.2 XP_006724689.1 15, 16 2261
          XM_011530883.1 XP_011529185.1 14, 15 2273
          XM_017029322.1 XP_016884811.1 8, 9 1216
          XM_017029323.1 XP_016884812.1 9, 10 1204
          XM_017029324.1 XP_016884813.1 8, 9 1182
          XM_017029325.1 XP_016884814.1 7, 8 2576
          XM_017029326.1 XP_016884815.1 7, 8 655
          XM_017029327.1 XP_016884816.1 6, 7 625
          GenBank mRNA AB033058.1 - 9, 10 1267
          AB209328.1 - 14, 15 2214
          AK303377.1 - 7, 8 726
          AK304020.1 - 9, 10 982
          AK315041.1 - 14, 15 2153
          AK316518.1 - 9, 10 968
          BC093864.1 - 14, 15 1915 7939709
          BC093866.1 - 14, 15 1915 7939711
          U49089.1 - 14, 15 2100

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of planar polarity
          chemical synaptic transmission
          nervous system development
          negative regulation of cell proliferation
          negative regulation of phosphatase activity
          ion transmembrane transport
          receptor clustering
          establishment or maintenance of epithelial cell apical/basal polarity
          GMP metabolic process
          GDP metabolic process
          receptor localization to synapse
          guanylate kinase activity
          protein binding
          protein C-terminus binding
          ligand-gated ion channel activity
          kinase binding
          phosphatase binding
          protein phosphatase binding
          PDZ domain binding
          ubiquitin protein ligase binding
          ionotropic glutamate receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0