Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ETV6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001987.4 | NP_001978.1 | 6 | 1303 | ||
XM_011520607.2 | XP_011518909.1 | 6 | 1553 | |||
XM_011520608.2 | XP_011518910.1 | 6 | 1892 | |||
XM_011520609.2 | XP_011518911.1 | 6 | 2167 | |||
XM_011520611.2 | XP_011518913.1 | 5 | 1100 | |||
XM_011520612.2 | XP_011518914.1 | 4 | 834 | |||
XM_017018990.1 | XP_016874479.1 | 5 | 1421 | |||
XM_017018991.1 | XP_016874480.1 | 10 | 2609 | |||
GenBank mRNA | AK289441.1 | - | 6 | 1171 | ||
BC043399.1 | - | 6 | 1303 | 6014394 | ||
KU178049.1 | - | 6 | 1029 | |||
U11732.1 | - | 6 | 1053 |