Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: F9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000133.3 | NP_000124.1 | 7 | 787 | ||
NM_001313913.1 | NP_001300842.1 | 6 | 673 | |||
XM_005262397.4 | XP_005262454.1 | 6 | 674 | |||
GenBank mRNA | A13997.1 | - | 7 | 759 | ||
AB186358.1 | - | 6 | 644 | |||
AK292749.1 | - | 7 | 758 | |||
BC109214.1 | - | 7 | 787 | 40007636 | ||
BC109215.1 | - | 7 | 787 | 40007637 | ||
FR846238.1 | - | 7 | 758 | |||
FR846239.1 | - | 7 | 758 | |||
FR846240.1 | - | 7 | 758 | |||
J00136.1 | - | 7 | 758 | |||
J00137.1 | - | 7 | 759 | |||
M11309.1 | - | 7 | 758 | |||
M35672.1 | - | 6 | 653 |