Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FANCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018115.2 | NP_001018125.1 | 11 | 925 | ||
NM_001319984.1 | NP_001306913.1 | 11 | 997 | |||
NM_033084.4 | NP_149075.2 | 11 | 925 | |||
GenBank mRNA | AF230336.1 | - | 11 | 868 | ||
AF340183.1 | - | 11 | 868 | |||
AK022613.1 | - | 4 | 327 | |||
AK307512.1 | - | 12 | 898 |