Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Instrument Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
Our Instagram
Our Facebook
Blog Behind the Bench
Customer Experience Center (CEC)
Ecommerce Exclusives
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FDFT1 SIRNA Assays ›
Gene Symbol:
FDFT1
Gene Name:
farnesyl-diphosphate farnesyltransferase 1
Gene Aliases:
DGPT, ERG9, SQS, SS
Chromosome Location:
Chr.8: 11795573 - 11839309 on Build GRCh38
UniGene ID:
Hs.593928
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS103617
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FDFT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001287742.1
NP_001274671.1
9
1613
NM_001287743.1
NP_001274672.1
8
1415
NM_001287744.1
NP_001274673.1
7
1242
NM_001287745.1
NP_001274674.1
8
1321
NM_001287747.1
NP_001274676.1
7
1278
NM_001287748.1
NP_001274677.1
7
940
NM_001287749.1
NP_001274678.1
7
1178
NM_001287750.1
NP_001274679.1
6
1180
NM_001287751.1
NP_001274680.1
6
1052
NM_001287756.1
NP_001274685.1
4
637
NM_004462.4
NP_004453.3
7
1124
XM_017013217.1
XP_016868706.1
9
1905
GenBank mRNA
AK057726.1
-
7
1242
AK098682.1
-
702
AK293545.1
-
7
1178
AK296043.1
-
6
867
AK297868.1
-
6
1123
AK300059.1
-
6
942
AK300245.1
-
7
735
AK301617.1
-
4
498
AK311246.1
-
6
891
AK315993.1
-
8
1191
AK316033.1
-
7
1145
AK316182.1
-
6
1140
AK316351.1
-
7
905
AK316531.1
-
7
890
AK316534.1
-
6
1052
BC003573.1
-
7
971
3538137
BC009251.2
-
7
971
2961623
BC029641.1
-
7
934
4514761
BC034440.1
-
7
934
BT006704.1
-
7
902
CR457033.1
-
7
902
L06070.1
-
7
911
L06105.1
-
7
946
S76822.1
-
7
967
X69141.1
-
7
993
Back To Top
More Information
Panther Classification:
Molecular Function -
transferase
Gene Ontology Categories:
Function(s)
Process(es)
steroid biosynthetic process
cholesterol biosynthetic process
isoprenoid biosynthetic process
farnesyl diphosphate metabolic process
oxidation-reduction process
farnesyl-diphosphate farnesyltransferase activity
oxidoreductase activity
squalene synthase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-4pmfl:80/100.66.77.34:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0