Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FMR1 SIRNA Assays ›
          Gene Symbol:
          FMR1
          Gene Name:
          fragile X mental retardation 1
          Gene Aliases:
          FMRP, FRAXA, POF, POF1
          Chromosome Location:
          Chr.X: 147911951 - 147951127 on Build GRCh38
          UniGene ID:
          Hs.103183
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS103776
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FMR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001185075.1 NP_001172004.1 4 464
          NM_001185076.1 NP_001172005.1 4 464
          NM_001185081.1 NP_001172010.1 4 464
          NM_001185082.1 NP_001172011.1 4 464
          NM_002024.5 NP_002015.1 4 464
          NR_033699.1 4 464
          NR_033700.1 4 464
          GenBank mRNA AB209188.1 - 1 689
          BC038998.1 - 4 429
          BC086957.1 - 4 494 30347992
          BQ213316.1 - 4 416
          KC774027.1 - 4 297
          KC774028.1 - 4 297
          KC774029.1 - 4 297
          KF856234.1 - 4 297
          KJ534836.1 - 4 235
          KJ534837.1 - 4 235
          KJ534838.1 - 4 235
          KM091457.1 - 4 297
          KM091458.1 - 4 297
          KM091459.1 - 4 297
          M67468.1 - 4 430
          S65791.1 - 4 454
          X69962.1 - 4 454

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_052804, XM_006229530, XM_006229531, XM_006229535, XM_008773597, and others.
          This siRNA also targets the following transcripts in Mouse: NM_008031, NM_001290424, AK053701, AK080948, AK053211, and others.

          Panther Classification:

          Molecular Function -

          translation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of alternative mRNA splicing, via spliceosome
          positive regulation of receptor internalization
          mRNA processing
          cellular response to DNA damage stimulus
          glutamate receptor signaling pathway
          RNA splicing
          negative regulation of translation
          gene silencing by RNA
          positive regulation of histone phosphorylation
          cellular response to UV
          regulation of mRNA stability
          modulation by host of viral RNA genome replication
          positive regulation of translation
          negative regulation of translational initiation
          regulation of neurotransmitter secretion
          mRNA transport
          regulation of filopodium assembly
          positive regulation of filopodium assembly
          regulation of gene silencing by miRNA
          regulation of dendritic spine development
          positive regulation of dendritic spine development
          cellular response to hydroxyurea
          cellular response to virus
          regulation of neuronal action potential
          negative regulation of long term synaptic depression
          positive regulation of intracellular transport of viral material
          negative regulation of voltage-gated calcium channel activity
          positive regulation of proteasomal protein catabolic process
          positive regulation of mRNA binding
          negative regulation of synaptic vesicle exocytosis
          positive regulation of gene silencing by miRNA
          negative regulation of cytoplasmic translation
          positive regulation of response to DNA damage stimulus
          G-quadruplex RNA binding
          chromatin binding
          RNA binding
          mRNA binding
          mRNA 3'-UTR binding
          protein binding
          microtubule binding
          poly(U) RNA binding
          translation repressor activity
          translation initiation factor binding
          RNA strand annealing activity
          poly(G) binding
          methylated histone binding
          siRNA binding
          miRNA binding
          RNA stem-loop binding
          identical protein binding
          protein homodimerization activity
          ribosome binding
          ion channel binding
          poly(A) RNA binding
          dynein binding
          protein heterodimerization activity
          mRNA 5'-UTR binding
          sequence-specific mRNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-6hb64:80/100.66.77.245:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0