Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FN1 SIRNA Assays ›
Gene Symbol:
FN1
Gene Name:
fibronectin 1
Gene Aliases:
CIG, ED-B, FINC, FN, FNZ, GFND, GFND2, LETS, MSF
Chromosome Location:
Chr.2: 215360440 - 215436167 on Build GRCh38
UniGene ID:
Hs.203717
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS103782
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001306129.1
NP_001293058.1
5
852
NM_001306130.1
NP_001293059.1
5
852
NM_001306131.1
NP_001293060.1
5
852
NM_001306132.1
NP_001293061.1
5
852
NM_002026.3
NP_002017.1
5
850
NM_054034.2
NP_473375.2
5
850
NM_212474.2
NP_997639.1
5
850
NM_212476.2
NP_997641.1
5
850
NM_212478.2
NP_997643.1
5
850
NM_212482.2
NP_997647.1
5
850
XM_005246397.1
XP_005246454.1
5
949
XM_005246398.1
XP_005246455.1
5
949
XM_005246399.1
XP_005246456.1
5
949
XM_005246401.1
XP_005246458.1
5
949
XM_005246402.1
XP_005246459.1
5
949
XM_005246403.1
XP_005246460.1
5
949
XM_005246404.1
XP_005246461.1
5
949
XM_005246405.1
XP_005246462.1
5
949
XM_005246406.1
XP_005246463.1
5
949
XM_005246407.1
XP_005246464.1
5
949
XM_005246408.1
XP_005246465.1
5
949
XM_005246409.1
XP_005246466.1
5
949
XM_005246410.1
XP_005246467.1
5
949
XM_005246411.1
XP_005246468.1
5
949
XM_005246412.1
XP_005246469.1
5
949
XM_005246414.1
XP_005246471.1
5
949
XM_005246416.1
XP_005246473.1
5
949
XM_017003692.1
XP_016859181.1
5
949
XM_017003693.1
XP_016859182.1
5
949
XM_017003694.1
XP_016859183.1
5
949
XM_017003695.1
XP_016859184.1
5
949
GenBank mRNA
AB191261.1
-
5
850
AB209840.1
-
5
842
AJ276395.1
-
5
640
AJ535086.1
-
5
640
AJ849445.1
-
5
640
AL832771.1
-
5
851
BC117176.1
-
5
830
40125727
BC143754.1
-
5
830
9052268
BC143763.1
-
5
830
9052277
BX537590.1
-
5
851
BX538017.1
-
5
851
BX538018.1
-
5
851
BX538045.1
-
5
890
BX640608.1
-
5
850
BX640731.1
-
5
851
BX640802.1
-
5
853
BX640803.1
-
5
850
BX640875.1
-
5
851
BX649182.1
-
5
853
CR749281.1
-
5
889
CR749316.1
-
5
849
CR749317.1
-
5
849
U42404.1
-
3
279
X02761.1
-
5
504
Back To Top
More Information
Panther Classification:
Molecular Function -
extracellular matrix structural protein
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
cell activation
regulation of protein phosphorylation
platelet degranulation
response to ischemia
acute-phase response
cell-substrate junction assembly
cell adhesion
calcium-independent cell-matrix adhesion
positive regulation of cell proliferation
glial cell migration
regulation of cell shape
response to wounding
response to ozone
positive regulation of gene expression
positive regulation of peptidase activity
peptide cross-linking
extracellular matrix disassembly
extracellular matrix organization
integrin activation
substrate adhesion-dependent cell spreading
cellular response to vascular endothelial growth factor stimulus
endodermal cell differentiation
cellular response to platelet-derived growth factor stimulus
wound healing
negative regulation of apoptotic process
positive regulation of axon extension
positive regulation of fibroblast proliferation
leukocyte migration
positive regulation of chemotaxis
response to glucocorticoid
regulation of ERK1 and ERK2 cascade
cellular response to lipopolysaccharide
cellular response to mercury ion
cellular response to glucose stimulus
cellular response to interleukin-1
cellular response to prostaglandin E stimulus
cellular response to transforming growth factor beta stimulus
cellular response to BMP stimulus
positive regulation of substrate-dependent cell migration, cell attachment to substrate
negative regulation of transforming growth factor-beta secretion
protease binding
integrin binding
protein binding
collagen binding
heparin binding
peptidase activator activity
identical protein binding
mercury ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-49ts5:80/100.66.72.225:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0