Search
Search

ADAM2| Gene Symbol: ADAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278113.1 | NP_001265042.1 | 17 | 1950 | ||
| NM_001278114.1 | NP_001265043.1 | 17 | 1818 | |||
| NM_001464.4 | NP_001455.3 | 18 | 2007 | |||
| XM_005273468.1 | XP_005273525.1 | 18 | 1855 | |||
| XM_011544479.1 | XP_011542781.1 | 14 | 1567 | |||
| XM_011544480.1 | XP_011542782.1 | 14 | 1477 | |||
| GenBank mRNA | AJ133005.1 | - | 18 | 1907 | ||
| AK301734.1 | - | 17 | 1756 | |||
| BC034957.2 | - | 17 | 1870 | 4826530 | ||
| BC064547.1 | - | 14 | 1460 | 5744846 | ||
| U38805.1 | - | 18 | 1971 | |||
| U52370.1 | - | 18 | 1910 | |||