Search
Search

GBP1| Gene Symbol: GBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002053.2 | NP_002044.2 | 2 | 319 | ||
| GenBank mRNA | AB208912.1 | - | 2 | 278 | ||
| AK291783.1 | - | 2 | 167 | |||
| AK298032.1 | - | 2 | 198 | |||
| AY920436.1 | - | 1 | 103 | |||
| BC002666.2 | - | 2 | 319 | 3606865 | ||
| BT006847.1 | - | 1 | 99 | |||
| M55542.1 | - | 2 | 167 | |||