This product has been added to your favorites list. Go to
My Favorites
LOADING 
- Gene Symbol:
-
GFI1
- Gene Name:
-
growth factor independent 1 transcriptional repressor
- Gene Aliases:
-
GFI-1, GFI1A, SCN2, ZNF163
- Chromosome Location:
-
Chr.1: 92473043 - 92486876 on Build GRCh38
- UniGene ID:
-
Hs.73172
- Species:
- Human
- Species Specific ID (Flybase ID):
-
-
siRNA Details
|
Gene Symbol:
GFI1 |
| Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
| RefSeq |
NM_001127215.1 |
NP_001120687.1 |
3, 4
|
466
|
|
| NM_001127216.1 |
NP_001120688.1 |
3, 4
|
459
|
|
| NM_005263.3 |
NP_005254.2 |
3, 4
|
552
|
|
| XM_005270749.3 |
XP_005270806.1 |
3, 4
|
908
|
|
| XM_011541245.2 |
XP_011539547.1 |
3, 4
|
1394
|
|
| XM_011541246.2 |
XP_011539548.1 |
3, 4
|
670
|
|
| GenBank mRNA |
AB209606.1 |
- |
2, 3
|
340
|
|
| AK123009.1 |
- |
3, 4
|
621
|
|
| AK313876.1 |
- |
3, 4
|
466
|
|
| BC032751.1 |
- |
3, 4
|
436
|
5585794 |
| BC074866.2 |
- |
2, 3
|
346
|
30915466 |
| BC074867.2 |
- |
2, 3
|
346
|
30915231 |
| DA927584.1 |
- |
1, 2
|
125
|
|
| DB129173.1 |
- |
3, 4
|
466
|
|
| DB137690.1 |
- |
3, 4
|
461
|
|
| U67369.1 |
- |
3, 4
|
569
|
|
Back To Top
Related Products
-
Anti-GAPDH, Mouse Monoclonal 6C5
-
Anti-β-Actin, Mouse Monoclonal AC-15
-
TaqMan® Gene Expression Cells-to-CT™
-
Neon™ Transfection System 100 µL Kit
-
Lipofectamine™ RNAiMAX Transfection Reagent
-
Invivofectamine 2.0
-
Lipofectamine™ 2000 Transfection Reagent
-
Neon™ Transfection System