Search
Search

GLS| Gene Symbol: GLS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256310.1 | NP_001243239.1 | 12 | 1577 | ||
| NM_014905.4 | NP_055720.3 | 12 | 1577 | |||
| XM_005246467.3 | XP_005246524.1 | 12 | 1577 | |||
| XM_006712435.1 | XP_006712498.1 | 12 | 1577 | |||
| GenBank mRNA | AB020645.1 | - | 12 | 1563 | ||
| AF097492.1 | - | 12 | 1211 | |||
| AF097493.1 | - | 5 | 305 | |||
| AF158555.1 | - | 13 | 1557 | |||
| AF223943.1 | - | 12 | 1332 | |||
| AF279697.1 | - | 9 | 662 | |||
| AF327434.1 | - | 12 | 1543 | |||
| AK001220.1 | - | 9 | 649 | |||
| AK289747.1 | - | 13 | 1553 | |||
| AK294927.1 | - | 1 | 101 | |||
| BC038507.1 | - | 13 | 1553 | 5263220 | ||
| CR749593.1 | - | 5 | 2036 | |||