Search
Search

GNAT2| Gene Symbol: GNAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005272.3 | NP_005263.1 | 5 | 728 | ||
| XM_011541264.2 | XP_011539566.1 | 6 | 819 | |||
| GenBank mRNA | AF493909.1 | - | 5 | 515 | ||
| BC000233.1 | - | 5 | 696 | 3350818 | ||
| BT009844.1 | - | 5 | 515 | |||