Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GPR31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005299.2 | NP_005290.2 | 1 | 544 | ||
GenBank mRNA | BC095537.1 | - | 1 | 541 | ||
CD246167.1 | - | 1 | 552 |