Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IL7R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002185.3 | NP_002176.2 | 8 | 1375 | ||
NR_120485.1 | 6 | 1112 | ||||
XM_005248299.3 | XP_005248356.1 | 7 | 1266 | |||
GenBank mRNA | AK315251.1 | - | 8 | 1274 | ||
BC067537.1 | - | 8 | 1352 | 6971840 | ||
BC067538.1 | - | 8 | 1352 | 6971842 | ||
BC067539.1 | - | 8 | 1352 | 6971844 | ||
BC067540.1 | - | 8 | 1352 | 6971846 | ||
BC069999.1 | - | 8 | 1352 | 6971843 | ||
FJ529208.1 | - | 6 | 1098 | |||
M29696.1 | - | 8 | 1294 |