Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IL15RA SIRNA Assays ›
Gene Symbol:
IL15RA
Gene Name:
interleukin 15 receptor subunit alpha
Gene Aliases:
CD215
Chromosome Location:
Chr.10: 5948897 - 5978741 on Build GRCh38
UniGene ID:
Hs.445124
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS105398
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IL15RA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243539.1
NP_001230468.1
4
815
NM_001256765.1
NP_001243694.1
5
764
NM_002189.3
NP_002180.1
4
611
NM_172200.2
NP_751950.2
3
512
NR_046362.1
2
521
XM_011519461.2
XP_011517763.1
6
1005
XM_011519462.2
XP_011517764.1
5
923
XM_011519463.2
XP_011517765.1
5
906
XM_011519464.2
XP_011517766.1
5
852
XM_011519465.1
XP_011517767.1
6
1005
XM_011519466.2
XP_011517768.1
6
1005
XM_011519467.2
XP_011517769.1
4
768
XM_011519468.1
XP_011517770.1
6
1005
XM_011519469.1
XP_011517771.1
5
906
XM_011519470.2
XP_011517772.1
3
671
XM_011519471.2
XP_011517773.1
4
736
XM_011519472.2
XP_011517774.1
3
575
XM_011519474.2
XP_011517776.1
3
558
XM_011519475.2
XP_011517777.1
2
476
XM_011519476.2
XP_011517778.1
2
466
XM_011519477.2
XP_011517779.1
2
471
XM_017016195.1
XP_016871684.1
5
852
XM_017016196.1
XP_016871685.1
4
577
XM_017016197.1
XP_016871686.1
5
592
XM_017016198.1
XP_016871687.1
4
736
XM_017016200.1
XP_016871689.1
3
1163
XM_017016201.1
XP_016871690.1
4
860
XM_017016202.1
XP_016871691.1
5
581
XM_017016203.1
XP_016871692.1
3
1163
GenBank mRNA
AF035279.1
-
4
546
AK304211.1
-
3
423
BC065506.1
-
2
513
BC074726.2
-
4
523
30915179
BC107777.1
-
4
679
6497994
BC121140.2
-
4
518
40120998
BC121141.2
-
4
523
40120999
BM546106.1
-
2
532
CR457064.1
-
3
384
CR542023.1
-
4
492
DC298692.1
-
3
517
HQ401283.1
-
5
645
JX987304.1
-
5
645
JX987305.1
-
5
645
JX987306.1
-
5
645
JX987307.1
-
5
645
JX987308.1
-
5
645
JX987309.1
-
5
645
JX987310.1
-
5
645
JX987311.1
-
5
645
U31628.1
-
4
574
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
cell proliferation
cytokine-mediated signaling pathway
signal transducer activity
cytokine receptor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-jmx4f:80/100.66.76.53:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0