Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IMPDH1 SIRNA Assays ›
          Gene Symbol:
          IMPDH1
          Gene Name:
          inosine monophosphate dehydrogenase 1
          Gene Aliases:
          IMPD, IMPD1, IMPDH-I, LCA11, RP10, sWSS2608
          Chromosome Location:
          Chr.7: 128392277 - 128409988 on Build GRCh38
          UniGene ID:
          Hs.654401
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS105421
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IMPDH1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000883.3 NP_000874.2 9, 10 965
          NM_001102605.1 NP_001096075.1 8, 9 935
          NM_001142573.1 NP_001136045.1 6, 7 735
          NM_001142574.1 NP_001136046.1 6, 7 720
          NM_001142575.1 NP_001136047.1 5, 6 660
          NM_001142576.1 NP_001136048.1 8, 9 866
          NM_001304521.1 NP_001291450.1 6, 7 763
          NM_183243.2 NP_899066.1 7, 8 857
          XM_017012168.1 XP_016867657.1 8, 9 935
          XM_017012169.1 XP_016867658.1 7, 8 857
          XM_017012170.1 XP_016867659.1 6, 7 764
          XM_017012171.1 XP_016867660.1 8, 9 1053
          XM_017012172.1 XP_016867661.1 8, 9 1053
          XM_017012173.1 XP_016867662.1 8, 9 935
          GenBank mRNA AK054640.1 - 5, 6 632
          AK054667.1 - 7, 8 857
          AK092452.1 - 8, 9 902
          AK122994.1 - 8, 9 934
          AK131294.1 - 6, 7 682
          AK293413.1 - 6, 7 692
          AK297943.1 - 6, 7 707
          BC033622.1 - 7, 8 848 5210847
          CD014008.1 - 7, 8 649
          CD014010.1 - 8, 9 771
          CR933672.1 - 901
          J05272.1 - 9, 10 1229

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          GMP biosynthetic process
          purine ribonucleoside monophosphate biosynthetic process
          response to cold
          lymphocyte proliferation
          oxidation-reduction process
          retina development in camera-type eye
          nucleotide binding
          nucleic acid binding
          DNA binding
          RNA binding
          IMP dehydrogenase activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0