Search
Search

ITGB2| Gene Symbol: ITGB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000211.4 | NP_000202.3 | 15, 16 | 2471 | ||
| NM_001127491.2 | NP_001120963.2 | 15, 16 | 2461 | |||
| NM_001303238.1 | NP_001290167.1 | 15, 16 | 2426 | |||
| XM_006724001.1 | XP_006724064.1 | 15, 16 | 2488 | |||
| XM_017028341.1 | XP_016883830.1 | 15, 16 | 2483 | |||
| GenBank mRNA | AB208909.1 | - | 14, 15 | 4294 | ||
| AK095992.1 | - | 15, 16 | 2426 | |||
| AK097216.1 | - | 6, 7 | 1638 | |||
| AK128344.1 | - | 15, 16 | 2296 | |||
| AK222505.1 | - | 15, 16 | 2328 | |||
| AK303488.1 | - | 15, 16 | 2339 | |||
| BC005861.2 | - | 14, 15 | 2282 | 3532902 | ||
| BC021077.1 | - | 14, 15 | 2205 | |||
| CA447686.1 | - | 1, 2 | 499 | |||
| L78790.1 | - | 14, 15 | 2118 | |||
| M15395.1 | - | 15, 16 | 2310 | |||
| Y00057.1 | - | 14, 15 | 2214 | |||