Search
Search

ITPKB| Gene Symbol: ITPKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002221.3 | NP_002212.3 | 4 | 2401 | ||
| GenBank mRNA | AJ242780.1 | - | 3 | 2118 | ||
| AK289700.1 | - | 3 | 847 | |||
| AK313801.1 | - | 3 | 866 | |||
| AY513281.1 | - | 4 | 642 | |||
| BQ434930.1 | - | 3 | 553 | |||
| X57206.1 | - | 3 | 740 | |||
| Y18024.2 | - | 3 | 2061 | |||