Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KCNMA1 SIRNA Assays ›
Gene Symbol:
KCNMA1
Gene Name:
potassium calcium-activated channel subfamily M alpha 1
Gene Aliases:
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
Chromosome Location:
Chr.10: 76869601 - 77637819 on Build GRCh38
UniGene ID:
Hs.144795
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS105761
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KCNMA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001014797.2
NP_001014797.1
28
3527
NM_001161352.1
NP_001154824.1
28
3689
NM_001161353.1
NP_001154825.1
28
3638
NM_001271518.1
NP_001258447.1
27
3365
NM_001271519.1
NP_001258448.1
29
3605
NM_001322829.1
NP_001309758.1
28
3524
NM_001322830.1
NP_001309759.1
30
3617
NM_001322832.1
NP_001309761.1
27
3515
NM_001322835.1
NP_001309764.1
29
3608
NM_001322836.1
NP_001309765.1
28
3524
NM_001322837.1
NP_001309766.1
29
3608
NM_001322838.1
NP_001309767.1
29
3562
NM_002247.3
NP_002238.2
27
3515
XM_005269776.3
XP_005269833.1
28
3585
XM_005269778.1
XP_005269835.1
29
3690
XM_005269781.2
XP_005269838.1
28
3610
XM_005269787.3
XP_005269844.1
27
3504
XM_005269789.2
XP_005269846.1
29
3529
XM_005269792.2
XP_005269849.1
28
3523
XM_005269796.2
XP_005269853.1
27
3436
XM_006717826.2
XP_006717889.1
28
3517
XM_011539773.1
XP_011538075.1
30
3822
XM_011539774.1
XP_011538076.1
31
3816
XM_011539775.1
XP_011538077.1
29
3810
XM_011539776.1
XP_011538078.1
29
3810
XM_011539777.1
XP_011538079.1
30
3771
XM_011539778.2
XP_011538080.1
29
3691
XM_011539779.1
XP_011538081.1
29
3723
XM_011539780.1
XP_011538082.1
29
3690
XM_011539781.2
XP_011538083.1
29
3648
XM_011539782.2
XP_011538084.1
28
3588
XM_011539783.1
XP_011538085.1
29
3603
XM_011539785.1
XP_011538087.1
29
3594
XM_017016207.1
XP_016871696.1
31
3903
XM_017016208.1
XP_016871697.1
31
3903
XM_017016209.1
XP_016871698.1
30
3891
XM_017016210.1
XP_016871699.1
30
3822
XM_017016211.1
XP_016871700.1
30
3735
XM_017016212.1
XP_016871701.1
30
3729
XM_017016213.1
XP_016871702.1
29
3723
XM_017016214.1
XP_016871703.1
29
3717
XM_017016215.1
XP_016871704.1
30
3684
XM_017016216.1
XP_016871705.1
29
3648
XM_017016217.1
XP_016871706.1
28
3588
XM_017016218.1
XP_016871707.1
28
3054
XM_017016222.1
XP_016871711.1
27
3504
XM_017016223.1
XP_016871712.1
26
2900
GenBank mRNA
AB113382.1
-
28
3454
AB113575.1
-
28
3389
AB209488.1
-
27
2934
AB524033.1
-
27
3188
AF025999.1
-
27
3266
AF118141.1
-
27
3143
AK310379.1
-
29
3562
AY040849.1
-
27
3280
BC137115.1
-
27
3418
9021112
BC137137.1
-
27
3418
9021134
BC144496.1
-
29
3508
9053023
BX648925.1
-
24
2680
U02632.1
-
21
2460
U11058.2
-
27
3229
U11717.1
-
27
3290
U13913.1
-
27
3416
U23767.1
-
27
3338
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
potassium ion transport
response to osmotic stress
cellular potassium ion homeostasis
response to carbon monoxide
regulation of ion transmembrane transport
regulation of membrane potential
positive regulation of apoptotic process
negative regulation of cell volume
response to calcium ion
micturition
smooth muscle contraction involved in micturition
potassium ion transmembrane transport
actin binding
voltage-gated potassium channel activity
protein binding
calcium-activated potassium channel activity
metal ion binding
large conductance calcium-activated potassium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0