Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KIF3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002254.6 | NP_002245.4 | 2 | 2241 | ||
XM_005264299.2 | XP_005264356.2 | 2 | 2241 | |||
GenBank mRNA | AF018164.1 | - | 2 | 1735 | ||
AF035621.1 | - | 2 | 1737 | |||
AK293738.1 | - | 3 | 1505 | |||
AK315324.1 | - | 2 | 2151 | |||
BC018932.2 | - | 2 | 93 | |||
BC042486.1 | - | 3 | 2148 | 3866587 | ||
BC092406.1 | - | 2 | 2126 | 6504352 | ||
BC130423.1 | - | 2 | 1812 | 40146454 | ||
BC132783.1 | - | 2 | 1812 | 40146805 | ||
BC144223.1 | - | 2 | 1812 | 9052743 | ||
BX571741.1 | - | 2 | 2164 |