Search
Search

LGALS4| Gene Symbol: LGALS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006149.3 | NP_006140.1 | 6 | 730 | ||
| XM_011526974.1 | XP_011525276.1 | 5 | 969 | |||
| GenBank mRNA | AB006781.1 | - | 6 | 572 | ||
| AF014838.1 | - | 6 | 529 | |||
| AK290269.1 | - | 6 | 575 | |||
| AK296194.1 | - | 5 | 599 | |||
| AK296604.1 | - | 5 | 663 | |||
| BC003661.2 | - | 6 | 559 | 2967411 | ||
| BC005146.2 | - | 6 | 559 | 2988384 | ||
| BC034750.1 | - | 6 | 555 | 5183145 | ||
| BT007032.1 | - | 6 | 516 | |||
| CR536544.1 | - | 6 | 516 | |||
| U82953.1 | - | 6 | 539 | |||