Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LRP1 SIRNA Assays ›
Gene Symbol:
LRP1
Gene Name:
LDL receptor related protein 1
Gene Aliases:
A2MR, APOER, APR, CD91, IGFBP3R, LRP, LRP1A, TGFBR5
Chromosome Location:
Chr.12: 57128401 - 57213377 on Build GRCh38
UniGene ID:
Hs.162757
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS106145
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LRP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002332.2
NP_002323.2
31, 32
5712
XM_017019303.1
XP_016874792.1
32, 33
5861
GenBank mRNA
X13916.1
-
31, 32
5712
Back To Top
More Information
Panther Classification:
Molecular Function -
apolipoprotein
Gene Ontology Categories:
Function(s)
Process(es)
retinoid metabolic process
receptor-mediated endocytosis
protein kinase C-activating G-protein coupled receptor signaling pathway
aging
cell proliferation
positive regulation of cholesterol efflux
negative regulation of neuron projection development
negative regulation of smooth muscle cell migration
cerebral cortex development
negative regulation of Wnt signaling pathway
positive regulation of lipid transport
regulation of cholesterol transport
regulation of phospholipase A2 activity
regulation of actin cytoskeleton organization
aorta morphogenesis
lipoprotein metabolic process
lipoprotein transport
apoptotic cell clearance
negative regulation of neuron apoptotic process
positive regulation of protein transport
beta-amyloid clearance
negative regulation of platelet-derived growth factor receptor-beta signaling pathway
protease binding
receptor activity
low-density lipoprotein receptor activity
calcium ion binding
protein binding
protein complex binding
apolipoprotein binding
lipoprotein transporter activity
poly(A) RNA binding
lipoprotein particle receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0