Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SMAD7 SIRNA Assays ›
          Gene Symbol:
          SMAD7
          Gene Name:
          SMAD family member 7
          Gene Aliases:
          CRCS3, MADH7, MADH8
          Chromosome Location:
          Chr.18: 48919853 - 48950711 on Build GRCh38
          UniGene ID:
          Hs.465087
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS106265
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SMAD7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001190821.1 NP_001177750.1 4 1134
          NM_001190822.1 NP_001177751.1 4 342
          NM_001190823.1 NP_001177752.1 2 435
          NM_005904.3 NP_005895.1 4 1137
          GenBank mRNA AF010193.1 - 4 1145
          AF015261.1 - 4 850
          AK131394.1 - 2 377
          AK301535.1 - 4 342
          BC074818.2 - 4 884 30915321
          BC074819.2 - 4 884 30915535
          BP363832.1 - 2 435

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          ureteric bud development
          transcription, DNA-templated
          transforming growth factor beta receptor signaling pathway
          regulation of epithelial to mesenchymal transition
          negative regulation of epithelial to mesenchymal transition
          negative regulation of peptidyl-threonine phosphorylation
          negative regulation of transcription by competitive promoter binding
          regulation of transforming growth factor beta receptor signaling pathway
          positive regulation of cell-cell adhesion
          negative regulation of cell migration
          BMP signaling pathway
          negative regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of BMP signaling pathway
          negative regulation of protein ubiquitination
          positive regulation of protein ubiquitination
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          regulation of activin receptor signaling pathway
          negative regulation of peptidyl-serine phosphorylation
          adherens junction assembly
          response to laminar fluid shear stress
          cellular protein complex localization
          intracellular signal transduction
          negative regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of transcription from RNA polymerase II promoter
          artery morphogenesis
          protein stabilization
          negative regulation of ubiquitin-protein transferase activity
          ventricular cardiac muscle tissue morphogenesis
          regulation of cardiac muscle contraction
          regulation of ventricular cardiac muscle cell membrane depolarization
          pathway-restricted SMAD protein phosphorylation
          negative regulation of pathway-restricted SMAD protein phosphorylation
          ventricular septum morphogenesis
          cellular response to transforming growth factor beta stimulus
          transcription factor activity, sequence-specific DNA binding
          protein binding
          collagen binding
          beta-catenin binding
          transforming growth factor beta receptor, inhibitory cytoplasmic mediator activity
          ubiquitin protein ligase binding
          type I transforming growth factor beta receptor binding
          transcription regulatory region DNA binding
          metal ion binding
          activin binding
          I-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0