Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308171.1 | NP_001295100.1 | 14 | 2111 | ||
NM_005925.2 | NP_005916.2 | 14 | 2111 | |||
XM_011526013.2 | XP_011524315.1 | 13 | 1877 | |||
XM_011526014.2 | XP_011524316.1 | 12 | 1724 | |||
GenBank mRNA | AY695931.1 | - | 14 | 2122 | ||
BC136559.1 | - | 14 | 2089 | 9020549 | ||
BC144244.1 | - | 14 | 2089 | 9052764 | ||
X81333.1 | - | 14 | 2110 |