Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MLH1 SIRNA Assays ›
          Gene Symbol:
          MLH1
          Gene Name:
          mutL homolog 1
          Gene Aliases:
          COCA2, FCC2, HNPCC, HNPCC2, hMLH1
          Chromosome Location:
          Chr.3: 36993350 - 37050846 on Build GRCh38
          UniGene ID:
          Hs.195364
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS106569
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MLH1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000249.3 NP_000240.1 11 1107
          NM_001167617.1 NP_001161089.1 11 902
          NM_001167618.1 NP_001161090.1 11 902
          NM_001167619.1 NP_001161091.1 10 815
          NM_001258271.1 NP_001245200.1 11 1107
          NM_001258273.1 NP_001245202.1 10 1016
          NM_001258274.1 NP_001245203.1 12 1052
          XM_005265161.1 XP_005265218.1 9 790
          XM_005265163.1 XP_005265220.1 11 907
          XM_005265164.1 XP_005265221.1 11 960
          XM_005265166.1 XP_005265223.1 10 821
          XM_017006449.1 XP_016861938.1 9 708
          XM_017006450.1 XP_016861939.1 9 753
          GenBank mRNA AB209848.1 - 11 885
          AK222810.1 - 11 914
          AK295359.1 - 11 881
          AK298324.1 - 11 853
          AK298583.1 - 11 875
          AK302807.1 - 13 836
          AK311365.1 - 11 856
          AK312609.1 - 11 923
          AK316074.1 - 10 788
          AK316264.1 - 10 833
          AY517558.1 - 11 907
          BC006850.1 - 11 959 3451538
          BX648844.1 - 12 1056
          DQ648890.1 - 3 196
          DQ648891.1 - 2 84
          EU188668.1 - 3 155
          EU188671.1 - 3 249
          U07343.1 - 11 930
          U07418.1 - 11 950

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA poly(A) tail shortening
          resolution of meiotic recombination intermediates
          mismatch repair
          double-strand break repair via nonhomologous end joining
          male meiosis chromosome segregation
          synapsis
          spermatogenesis
          intrinsic apoptotic signaling pathway in response to DNA damage
          female meiosis chromosome segregation
          somatic hypermutation of immunoglobulin genes
          meiotic metaphase I plate congression
          meiotic telomere clustering
          isotype switching
          negative regulation of mitotic recombination
          oogenesis
          meiotic spindle midzone assembly
          chromatin binding
          single-stranded DNA binding
          protein binding
          ATP binding
          ATPase activity
          guanine/thymine mispair binding
          MutSalpha complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0