Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEPT2 SIRNA Assays ›
Gene Symbol:
SEPT2
Gene Name:
septin 2
Gene Aliases:
DIFF6, NEDD-5, NEDD5, Pnutl3, hNedd5
Chromosome Location:
Chr.2: 241315187 - 241354027 on Build GRCh38
UniGene ID:
Hs.721234
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS107066
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEPT2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001008491.2
NP_001008491.1
6
786
NM_001008492.2
NP_001008492.1
6
468
NM_001282972.1
NP_001269901.1
6
517
NM_001282973.1
NP_001269902.1
9
741
NM_001321029.1
NP_001307958.1
6
502
NM_001321030.1
NP_001307959.1
6
500
NM_001321031.1
NP_001307960.1
7
603
NM_001321032.1
NP_001307961.1
5
383
NM_001321033.1
NP_001307962.1
7
619
NM_001321034.1
NP_001307963.1
7
618
NM_001321035.1
NP_001307964.1
6
484
NM_004404.4
NP_004395.1
5
383
NM_006155.2
NP_006146.1
5
685
XM_011511237.1
XP_011509539.1
5
349
XM_011511238.1
XP_011509540.1
4
217
XM_011511240.1
XP_011509542.1
4
226
XM_017004189.1
XP_016859678.1
7
578
XM_017004190.1
XP_016859679.1
7
513
XM_017004191.1
XP_016859680.1
7
578
XM_017004192.1
XP_016859681.1
7
578
XM_017004193.1
XP_016859682.1
5
383
XM_017004194.1
XP_016859683.1
7
533
XM_017004195.1
XP_016859684.1
7
737
XM_017004196.1
XP_016859685.1
7
619
XM_017004197.1
XP_016859686.1
6
434
XM_017004198.1
XP_016859687.1
6
496
XM_017004199.1
XP_016859688.1
7
535
XM_017004200.1
XP_016859689.1
6
498
XM_017004201.1
XP_016859690.1
6
620
XM_017004202.1
XP_016859691.1
5
602
XM_017004203.1
XP_016859692.1
6
426
XM_017004204.1
XP_016859693.1
5
341
XM_017004205.1
XP_016859694.1
7
533
XM_017004206.1
XP_016859695.1
6
434
XM_017004207.1
XP_016859696.1
3
235
XM_017004208.1
XP_016859697.1
6
445
XM_017004209.1
XP_016859698.1
5
332
XM_017004210.1
XP_016859699.1
5
376
XM_017004211.1
XP_016859700.1
4
229
XM_017004212.1
XP_016859701.1
4
481
XM_017004213.1
XP_016859702.1
7
578
GenBank mRNA
AF038404.1
-
5
322
AK294563.1
-
6
425
BC014455.1
-
6
665
4906575
BC033559.1
-
9
719
4821654
BG721140.1
-
5
385
BI561073.1
-
5
599
BX648000.1
-
6
402
D28540.1
-
4
306
D63878.1
-
5
502
DA544741.1
-
6
487
DB091579.1
-
5
383
DB277926.1
-
7
519
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of L-glutamate transport
mitotic nuclear division
smoothened signaling pathway
neuron projection development
regulation of protein localization
cilium assembly
regulation of catalytic activity
cell division
cell-cell adhesion
protein binding
GTP binding
enzyme regulator activity
protein complex scaffold
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-dh5dw:80/100.66.76.134:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0