Search
Search

NPR1| Gene Symbol: NPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000906.3 | NP_000897.3 | 5, 6 | 1686 | ||
| XM_005245218.1 | XP_005245275.1 | 5, 6 | 1757 | |||
| XM_017001374.1 | XP_016856863.1 | 5, 6 | 1757 | |||
| GenBank mRNA | AK025024.1 | - | 5, 6 | 1686 | ||
| AK300446.1 | - | 4, 5 | 892 | |||
| BC063304.1 | - | 5, 6 | 1735 | 30331757 | ||
| X15357.1 | - | 5, 6 | 1308 | |||