Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: OSBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002556.2 | NP_002547.1 | 8 | 1990 | ||
GenBank mRNA | AF185696.1 | - | 8 | 1990 | ||
AK299071.1 | - | 4 | 434 | |||
AK310538.1 | - | 3 | 369 | |||
BC011581.1 | - | 8 | 1578 | 4560111 | ||
BC063121.1 | - | 2 | 349 | |||
M86917.1 | - | 8 | 1990 |