Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PDE8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243137.1 | NP_001230066.1 | 16 | 1437 | ||
NM_002605.2 | NP_002596.1 | 16 | 1639 | |||
NM_173454.1 | NP_775656.1 | 15 | 1501 | |||
XM_011521668.2 | XP_011519970.1 | 9 | 1773 | |||
XM_017022308.1 | XP_016877797.1 | 13 | 1870 | |||
XM_017022309.1 | XP_016877798.1 | 14 | 1237 | |||
XM_017022310.1 | XP_016877799.1 | 16 | 2176 | |||
XM_017022311.1 | XP_016877800.1 | 17 | 2374 | |||
GenBank mRNA | AF056490.1 | - | 14 | 1102 | ||
AF332653.1 | - | 16 | 1586 | |||
AF388183.1 | - | 16 | 1632 | |||
AF388184.1 | - | 15 | 1494 | |||
AF388185.1 | - | 14 | 1415 | |||
AF388186.1 | - | 15 | 1553 | |||
AF388187.1 | - | 17 | 1830 | |||
AK074280.1 | - | 16 | 1437 | |||
AK127232.1 | - | 13 | 2920 | |||
BC060762.1 | - | 17 | 1702 | 30344110 | ||
BC075822.1 | - | 16 | 1530 | 6296489 |