Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE9A SIRNA Assays ›
Gene Symbol:
PDE9A
Gene Name:
phosphodiesterase 9A
Gene Aliases:
HSPDE9A2
Chromosome Location:
Chr.21: 42653752 - 42775509 on Build GRCh38
UniGene ID:
Hs.473927
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS107744
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE9A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001567.1
NP_001001567.1
6
437
NM_001001568.1
NP_001001568.1
4
288
NM_001001569.1
NP_001001569.1
3
233
NM_001001570.1
NP_001001570.1
6
458
NM_001001571.1
NP_001001571.1
5
366
NM_001001572.1
NP_001001572.1
3
244
NM_001001573.1
NP_001001573.1
4
424
NM_001001574.1
NP_001001574.1
5
359
NM_001001575.1
NP_001001575.1
2
189
NM_001001576.1
NP_001001576.1
3
233
NM_001001577.1
NP_001001577.1
3
267
NM_001001578.1
NP_001001578.1
4
311
NM_001001579.1
NP_001001579.1
4
304
NM_001001580.1
NP_001001580.1
2
189
NM_001001581.1
NP_001001581.1
5
468
NM_001001582.1
NP_001001582.1
6
546
NM_001001583.1
NP_001001583.1
6
539
NM_001001584.2
NP_001001584.1
5
367
NM_001001585.1
NP_001001585.1
5
491
NM_001315533.1
NP_001302462.1
7
513
NM_002606.2
NP_002597.1
7
617
XM_011529598.2
XP_011527900.1
7
539
XM_011529600.2
XP_011527902.1
7
590
XM_017028366.1
XP_016883855.1
5
355
XM_017028367.1
XP_016883856.1
7
590
GenBank mRNA
AF048837.1
-
7
614
AF067223.1
-
7
566
AF067224.1
-
6
385
AF067225.1
-
4
227
AF067226.1
-
3
172
AK001165.1
-
4
219
AK001307.1
-
6
408
AK294112.1
-
4
254
AK314679.1
-
7
590
AY196299.1
-
6
431
AY196300.1
-
5
339
AY196301.1
-
3
217
AY196302.1
-
4
397
AY196303.1
-
5
332
AY196304.1
-
2
162
AY196305.1
-
3
206
AY196306.1
-
3
240
AY196307.1
-
4
284
AY196308.1
-
4
277
AY196309.1
-
2
162
AY196310.1
-
5
441
AY196311.1
-
6
519
AY196312.1
-
6
512
AY196313.1
-
5
339
AY196314.1
-
5
464
AY242121.1
-
5
303
AY701187.1
-
7
510
BC009047.1
-
6
411
3874635
BT007016.1
-
6
374
KU178246.1
-
6
374
KU178247.1
-
7
554
KU178248.1
-
3
170
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphodiesterase
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
positive regulation of cardiac muscle hypertrophy
cGMP-mediated signaling
cGMP metabolic process
cGMP catabolic process
3',5'-cyclic-nucleotide phosphodiesterase activity
protein binding
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-nkbdw:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline