Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PECAM1 SIRNA Assays ›
Gene Symbol:
PECAM1
Gene Name:
platelet and endothelial cell adhesion molecule 1
Gene Aliases:
CD31, CD31/EndoCAM, GPIIA', PECA1, PECAM-1, endoCAM
Chromosome Location:
Chr.17: 64319415 - 64413844 on Build GRCh38
UniGene ID:
Hs.376675
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS107804
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PECAM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000442.4
NP_000433.4
8
1802
XM_005276880.1
XP_005276937.1
8
2088
XM_005276881.1
XP_005276938.1
8
2088
XM_005276882.1
XP_005276939.1
8
2088
XM_005276883.2
XP_005276940.1
8
2088
XM_011524889.2
XP_011523191.1
9
1818
XM_011524890.1
XP_011523192.1
9
1767
XM_017024738.1
XP_016880227.1
9
1711
XM_017024739.1
XP_016880228.1
8
2088
XM_017024740.1
XP_016880229.1
9
1781
XM_017024741.1
XP_016880230.1
8
2088
GenBank mRNA
AF281287.1
-
8
1672
AF281288.1
-
8
1672
AF281289.1
-
8
1672
AF281290.1
-
8
1672
AF281291.1
-
8
1672
AF281292.1
-
8
1672
AF281293.1
-
8
1672
AF281294.1
-
8
1672
AF281295.1
-
8
1672
AF281296.1
-
8
1672
AF281297.1
-
8
1672
AF281298.1
-
8
1672
AF281299.1
-
8
1672
AF281300.1
-
8
1672
AF281301.1
-
8
1672
AF393676.1
-
8
1672
AF393677.1
-
8
1672
AF393678.1
-
8
1672
AK290692.1
-
8
1605
AK303959.1
-
3
576
AK304166.1
-
7
1602
BC022512.1
-
8
1782
4816878
BC051822.1
-
8
1802
5768579
JQ287500.1
-
8
1589
M28526.1
-
8
1730
M37780.1
-
8
1714
S66450.1
-
1
71
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin receptor superfamily
Gene Ontology Categories:
Function(s)
Process(es)
platelet degranulation
phagocytosis
cell adhesion
signal transduction
cell recognition
extracellular matrix organization
leukocyte migration
diapedesis
glomerular endothelium development
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-t9swc:80/100.66.76.85:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0