Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PEX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000287.3 | NP_000278.3 | 4 | 1310 | ||
NM_001316313.1 | NP_001303242.1 | 4 | 1046 | |||
NR_133009.1 | 4 | 1310 | ||||
XM_011514661.1 | XP_011512963.1 | 3 | 1226 | |||
GenBank mRNA | AB051076.1 | - | 4 | 1217 | ||
AB051077.1 | - | 4 | 953 | |||
AB051078.1 | - | 4 | 1217 | |||
AB208906.1 | - | 4 | 1270 | |||
AK225704.1 | - | 4 | 1248 | |||
AK314237.1 | - | 4 | 1246 | |||
BC033487.1 | - | 4 | 1225 | |||
BC048331.1 | - | 4 | 1233 | 5189685 | ||
D83703.1 | - | 4 | 1287 |