Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPOX SIRNA Assays ›
Gene Symbol:
PPOX
Gene Name:
protoporphyrinogen oxidase
Gene Aliases:
PPO, V290M, VP
Chromosome Location:
Chr.1: 161165728 - 161178277 on Build GRCh38
UniGene ID:
Hs.517373
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS108337
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
459.00
Your Price
Online offer:
459.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPOX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000309.3
NP_000300.1
10
1272
NM_001122764.1
NP_001116236.1
10
1333
XM_005245291.3
XP_005245348.2
10
1311
XM_005245295.3
XP_005245352.2
9
1159
XM_006711402.2
XP_006711465.2
10
1278
XM_006711403.2
XP_006711466.2
10
1267
XM_006711404.3
XP_006711467.2
10
1215
XM_006711406.2
XP_006711469.2
10
1188
XM_011509663.2
XP_011507965.1
10
1398
XM_011509664.1
XP_011507966.1
10
1169
XM_011509665.2
XP_011507967.1
10
1396
XM_011509667.2
XP_011507969.1
10
1339
XM_011509668.2
XP_011507970.1
10
1139
XM_011509669.1
XP_011507971.1
10
1152
XM_011509670.2
XP_011507972.1
9
1287
XM_011509671.1
XP_011507973.1
10
1169
XM_011509672.2
XP_011507974.1
9
1278
XM_011509673.2
XP_011507975.1
8
1135
XM_011509675.2
XP_011507977.1
9
1220
XM_011509676.1
XP_011507978.1
9
1483
XM_011509677.1
XP_011507979.1
9
1416
XM_011509678.1
XP_011507980.1
10
1376
XM_011509679.1
XP_011507981.1
10
1184
XM_011509681.1
XP_011507983.1
9
1101
XM_011509682.1
XP_011507984.1
9
1111
XM_017001561.1
XP_016857050.1
9
1804
XM_017001562.1
XP_016857051.1
10
1257
XM_017001563.1
XP_016857052.1
10
1355
XM_017001564.1
XP_016857053.1
6
873
XM_017001565.1
XP_016857054.1
10
1257
XM_017001566.1
XP_016857055.1
10
1366
XM_017001567.1
XP_016857056.1
9
1159
XM_017001568.1
XP_016857057.1
10
1347
XM_017001569.1
XP_016857058.1
9
1101
XM_017001570.1
XP_016857059.1
7
902
GenBank mRNA
AK094855.1
-
9
1105
AK298912.1
-
4
448
AK302298.1
-
9
1143
BC002357.2
-
10
1253
2821983
D38537.1
-
10
1320
U26446.1
-
10
1234
Back To Top
More Information
Panther Classification:
Molecular Function -
oxidase
Gene Ontology Categories:
Function(s)
Process(es)
porphyrin-containing compound biosynthetic process
protoporphyrinogen IX biosynthetic process
heme biosynthetic process
response to drug
oxidation-reduction process
oxygen-dependent protoporphyrinogen oxidase activity
flavin adenine dinucleotide binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0