Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPP2R5C SIRNA Assays ›
          Gene Symbol:
          PPP2R5C
          Gene Name:
          protein phosphatase 2 regulatory subunit B'gamma
          Gene Aliases:
          B56G, PR61G
          Chromosome Location:
          Chr.14: 101760573 - 101927992 on Build GRCh38
          UniGene ID:
          Hs.368264
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS108389
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPP2R5C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001161725.1 NP_001155197.1 5, 6 592
          NM_001161726.1 NP_001155198.1 5, 6 664
          NM_002719.3 NP_002710.2 3, 4 543
          NM_178586.2 NP_848701.1 3, 4 543
          NM_178587.2 NP_848702.1 3, 4 543
          XM_005267819.1 XP_005267876.1 5, 6 653
          XM_005267820.1 XP_005267877.1 5, 6 593
          XM_005267822.3 XP_005267879.1 5, 6 653
          XM_005267823.1 XP_005267880.1 5, 6 653
          XM_005267824.1 XP_005267881.1 5, 6 653
          XM_005267826.1 XP_005267883.1 3, 4 366
          XM_005267827.1 XP_005267884.1 5, 6 588
          XM_011536914.2 XP_011535216.1 4, 5 457
          XM_011536915.2 XP_011535217.1 4, 5 540
          XM_011536916.2 XP_011535218.1 4, 5 1539
          XM_011536918.1 XP_011535220.1 4, 5 694
          XM_011536919.2 XP_011535221.1 4, 5 676
          XM_011536920.1 XP_011535222.1 4, 5 570
          XM_011536921.2 XP_011535223.1 3, 4 459
          XM_017021422.1 XP_016876911.1 4, 5 609
          XM_017021423.1 XP_016876912.1 3, 4 543
          XM_017021424.1 XP_016876913.1 4, 5 570
          GenBank mRNA AB451341.1 - 3, 4 403
          AK131391.1 - 5, 6 577
          AK302470.1 - 5, 6 492
          AK315072.1 - 3, 4 479
          BC016183.1 - 3, 4 441 4855950
          BI560029.1 - 3, 4 517
          BX248254.1 - 1, 2 38
          D26445.1 - 3, 4 400
          DA853338.1 - 3, 4 543
          L42375.1 - 3, 4 373
          U37352.1 - 3, 4 461
          Z69030.1 - 3, 4 481

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          negative regulation of cell proliferation
          regulation of phosphatidylinositol 3-kinase signaling
          regulation of protein phosphatase type 2A activity
          proteasome-mediated ubiquitin-dependent protein catabolic process
          protein binding
          protein phosphatase type 2A regulator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nq57x:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0