Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPP2R5C SIRNA Assays ›
Gene Symbol:
PPP2R5C
Gene Name:
protein phosphatase 2 regulatory subunit B'gamma
Gene Aliases:
B56G, PR61G
Chromosome Location:
Chr.14: 101760573 - 101927992 on Build GRCh38
UniGene ID:
Hs.368264
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS108390
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPP2R5C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001161725.1
NP_001155197.1
6, 7
677
NM_001161726.1
NP_001155198.1
6, 7
749
NM_002719.3
NP_002710.2
4, 5
628
NM_178586.2
NP_848701.1
4, 5
628
NM_178587.2
NP_848702.1
4, 5
628
XM_005267819.1
XP_005267876.1
6, 7
738
XM_005267820.1
XP_005267877.1
6, 7
678
XM_005267822.3
XP_005267879.1
6, 7
738
XM_005267823.1
XP_005267880.1
6, 7
738
XM_005267824.1
XP_005267881.1
6, 7
738
XM_005267826.1
XP_005267883.1
4, 5
451
XM_005267827.1
XP_005267884.1
6, 7
673
XM_011536914.2
XP_011535216.1
5, 6
542
XM_011536915.2
XP_011535217.1
5, 6
625
XM_011536916.2
XP_011535218.1
5, 6
1624
XM_011536918.1
XP_011535220.1
5, 6
779
XM_011536919.2
XP_011535221.1
5, 6
761
XM_011536920.1
XP_011535222.1
5, 6
655
XM_011536921.2
XP_011535223.1
4, 5
544
XM_017021422.1
XP_016876911.1
5, 6
694
XM_017021423.1
XP_016876912.1
4, 5
628
XM_017021424.1
XP_016876913.1
5, 6
655
GenBank mRNA
AB451341.1
-
4, 5
488
AK131391.1
-
6, 7
662
AK302470.1
-
6, 7
577
AK315072.1
-
4, 5
564
BC016183.1
-
4, 5
526
4855950
BX248254.1
-
2, 3
123
D26445.1
-
4, 5
485
L42375.1
-
4, 5
458
U37352.1
-
4, 5
546
Z69030.1
-
4, 5
566
Back To Top
More Information
Panther Classification:
Molecular Function -
protein phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
negative regulation of cell proliferation
regulation of phosphatidylinositol 3-kinase signaling
regulation of protein phosphatase type 2A activity
proteasome-mediated ubiquitin-dependent protein catabolic process
protein binding
protein phosphatase type 2A regulator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-qwrcb:80/100.66.75.127:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0