Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HTRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002775.4 | NP_002766.1 | 3 | 738 | ||
GenBank mRNA | AF070555.1 | - | 1 | 13 | ||
AF097709.1 | - | 3 | 181 | |||
AK091944.1 | - | 5 | 513 | |||
AK092476.1 | - | 4 | 519 | |||
AK290089.1 | - | 3 | 229 | |||
BC011352.1 | - | 4 | 505 | |||
BC031082.1 | - | 3 | 646 | |||
D87258.1 | - | 3 | 658 | |||
DA633502.1 | - | 5 | 513 | |||
Y07921.1 | - | 3 | 648 |