Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTCH1 SIRNA Assays ›
          Gene Symbol:
          PTCH1
          Gene Name:
          patched 1
          Gene Aliases:
          BCNS, HPE7, NBCCS, PTC, PTC1, PTCH, PTCH11
          Chromosome Location:
          Chr.9: 95442982 - 95516965 on Build GRCh38
          UniGene ID:
          Hs.494538
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS108758
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTCH1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000264.3 NP_000255.2 9 1425
          NM_001083602.1 NP_001077071.1 9 1533
          NM_001083603.1 NP_001077072.1 9 1379
          NM_001083604.1 NP_001077073.1 9 1143
          NM_001083605.1 NP_001077074.1 9 1126
          NM_001083606.1 NP_001077075.1 9 1225
          NM_001083607.1 NP_001077076.1 9 1299
          GenBank mRNA AB209495.1 - 6 874
          AB233422.1 - 4 377
          U43148.1 - 9 1225
          U59464.1 - 9 1237

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          branching involved in ureteric bud morphogenesis
          in utero embryonic development
          cell fate determination
          neural tube closure
          heart morphogenesis
          smoothened signaling pathway
          regulation of mitotic cell cycle
          brain development
          regulation of smoothened signaling pathway
          response to mechanical stimulus
          organ morphogenesis
          dorsal/ventral pattern formation
          epidermal cell fate specification
          response to chlorate
          positive regulation of cholesterol efflux
          protein processing
          spinal cord motor neuron differentiation
          neural tube patterning
          neural plate axis specification
          embryonic limb morphogenesis
          response to estradiol
          response to retinoic acid
          regulation of protein localization
          limb morphogenesis
          hindlimb morphogenesis
          negative regulation of multicellular organism growth
          response to drug
          glucose homeostasis
          negative regulation of sequence-specific DNA binding transcription factor activity
          keratinocyte proliferation
          positive regulation of epidermal cell differentiation
          negative regulation of osteoblast differentiation
          negative regulation of smoothened signaling pathway
          positive regulation of transcription, DNA-templated
          embryonic organ development
          negative regulation of epithelial cell proliferation
          negative regulation of cell division
          pharyngeal system development
          mammary gland duct morphogenesis
          mammary gland epithelial cell differentiation
          smoothened signaling pathway involved in dorsal/ventral neural tube patterning
          cell differentiation involved in kidney development
          somite development
          cellular response to cholesterol
          renal system development
          cell proliferation involved in metanephros development
          protein targeting to plasma membrane
          patched binding
          smoothened binding
          protein binding
          hedgehog receptor activity
          heparin binding
          cholesterol binding
          cyclin binding
          protein complex binding
          hedgehog family protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0