Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PEX5 SIRNA Assays ›
Gene Symbol:
PEX5
Gene Name:
peroxisomal biogenesis factor 5
Gene Aliases:
PBD2A, PBD2B, PTS1-BP, PTS1R, PXR1, RCDP5
Chromosome Location:
Chr.12: 7188072 - 7218574 on Build GRCh38
UniGene ID:
Hs.567327
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS108928
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
459.00
Your Price
Online offer:
459.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PEX5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000319.4
NP_000310.2
11
1139
NM_001131023.1
NP_001124495.1
12
1273
NM_001131024.1
NP_001124496.1
11
1117
NM_001131025.1
NP_001124497.1
12
1694
NM_001131026.1
NP_001124498.1
13
1727
NM_001300789.1
NP_001287718.1
12
1299
XM_005253455.1
XP_005253512.1
11
1183
XM_011520793.2
XP_011519095.1
12
1288
XM_011520795.1
XP_011519097.1
12
1705
XM_011520797.1
XP_011519099.1
12
1198
XM_011520798.1
XP_011519100.1
13
1407
XM_011520799.2
XP_011519101.1
12
1847
XM_011520800.1
XP_011519102.1
12
1293
XM_011520801.2
XP_011519103.1
11
1176
XM_011520802.2
XP_011519104.1
11
1050
XM_017019744.1
XP_016875233.1
11
1632
XM_017019745.1
XP_016875234.1
11
1633
XM_017019746.1
XP_016875235.1
10
1521
XM_017019747.1
XP_016875236.1
10
1500
XM_017019748.1
XP_016875237.1
12
1896
XM_017019749.1
XP_016875238.1
12
1660
XM_017019750.1
XP_016875239.1
12
1174
XM_017019751.1
XP_016875240.1
11
1198
XM_017019752.1
XP_016875241.1
12
1296
XM_017019753.1
XP_016875242.1
11
1182
XM_017019754.1
XP_016875243.1
11
1393
XM_017019755.1
XP_016875244.1
11
1791
XM_017019756.1
XP_016875245.1
11
1063
XM_017019757.1
XP_016875246.1
12
1272
GenBank mRNA
AK225126.1
-
11
1072
AK292256.1
-
13
1727
AK299105.1
-
11
1117
AK299534.1
-
7
628
AK301700.1
-
8
833
AK302742.1
-
12
1233
AK303515.1
-
12
1299
AK312317.1
-
12
1632
AK316250.1
-
11
1072
BC010621.2
-
12
1162
3872822
KU178292.1
-
11
1123
KU178293.1
-
11
1147
KU178294.1
-
10
1036
U19721.1
-
10
1050
X84899.1
-
11
1165
Z48054.1
-
11
1122
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
protein import into peroxisome matrix
protein import into peroxisome matrix, docking
protein import into peroxisome membrane
negative regulation of protein homotetramerization
peroxisome targeting sequence binding
peroxisome matrix targeting signal-1 binding
protein binding
enzyme binding
small GTPase binding
protein N-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0