Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PEX5 SIRNA Assays ›
Gene Symbol:
PEX5
Gene Name:
peroxisomal biogenesis factor 5
Gene Aliases:
PBD2A, PBD2B, PTS1-BP, PTS1R, PXR1, RCDP5
Chromosome Location:
Chr.12: 7188072 - 7218574 on Build GRCh38
UniGene ID:
Hs.567327
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS108929
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PEX5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000319.4
NP_000310.2
11, 12
1174
NM_001131023.1
NP_001124495.1
12, 13
1308
NM_001131024.1
NP_001124496.1
11, 12
1152
NM_001131025.1
NP_001124497.1
12, 13
1729
NM_001131026.1
NP_001124498.1
13, 14
1762
NM_001300789.1
NP_001287718.1
12, 13
1334
XM_005253455.1
XP_005253512.1
11, 12
1218
XM_011520793.2
XP_011519095.1
12, 13
1323
XM_011520795.1
XP_011519097.1
12, 13
1740
XM_011520797.1
XP_011519099.1
12, 13
1233
XM_011520798.1
XP_011519100.1
13, 14
1442
XM_011520799.2
XP_011519101.1
12, 13
1882
XM_011520800.1
XP_011519102.1
12, 13
1328
XM_011520801.2
XP_011519103.1
11, 12
1211
XM_011520802.2
XP_011519104.1
11, 12
1085
XM_017019744.1
XP_016875233.1
11, 12
1667
XM_017019745.1
XP_016875234.1
11, 12
1668
XM_017019746.1
XP_016875235.1
10, 11
1556
XM_017019747.1
XP_016875236.1
10, 11
1535
XM_017019748.1
XP_016875237.1
12, 13
1931
XM_017019749.1
XP_016875238.1
12, 13
1695
XM_017019750.1
XP_016875239.1
12, 13
1209
XM_017019751.1
XP_016875240.1
11, 12
1233
XM_017019752.1
XP_016875241.1
12, 13
1331
XM_017019753.1
XP_016875242.1
11, 12
1217
XM_017019754.1
XP_016875243.1
11, 12
1428
XM_017019755.1
XP_016875244.1
11, 12
1826
XM_017019756.1
XP_016875245.1
11, 12
1098
XM_017019757.1
XP_016875246.1
12, 13
1307
GenBank mRNA
AK225126.1
-
11, 12
1107
AK292256.1
-
13, 14
1762
AK299105.1
-
11, 12
1152
AK299534.1
-
7, 8
663
AK301700.1
-
8, 9
868
AK302742.1
-
12, 13
1268
AK303515.1
-
12, 13
1334
AK312317.1
-
12, 13
1667
AK316250.1
-
11, 12
1107
BC010621.2
-
12, 13
1197
3872822
KU178292.1
-
11, 12
1158
KU178293.1
-
11, 12
1182
KU178294.1
-
10, 11
1071
U19721.1
-
10, 11
1085
X84899.1
-
11, 12
1200
Z48054.1
-
11, 12
1157
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
protein import into peroxisome matrix
protein import into peroxisome matrix, docking
protein import into peroxisome membrane
negative regulation of protein homotetramerization
peroxisome targeting sequence binding
peroxisome matrix targeting signal-1 binding
protein binding
enzyme binding
small GTPase binding
protein N-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-mc8hj:80/100.66.75.242:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0