Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RBBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198719.1 | NP_001185648.1 | 6 | 1058 | ||
NM_002893.3 | NP_002884.1 | 6 | 1070 | |||
GenBank mRNA | AK091911.1 | - | 6 | 1055 | ||
AK291567.1 | - | 6 | 814 | |||
AK310309.1 | - | 3 | 359 | |||
BC114500.1 | - | 6 | 968 | 40083852 | ||
BC114501.1 | - | 6 | 968 | 40083851 | ||
BQ775345.1 | - | 4 | 654 | |||
CR541701.1 | - | 6 | 712 | |||
U35143.1 | - | 6 | 973 | |||
X72841.1 | - | 6 | 997 |