Search
Search

RELB| Gene Symbol: RELB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006509.3 | NP_006500.2 | 8, 9 | 1132 | ||
| XM_005259127.3 | XP_005259184.1 | 7, 8 | 1102 | |||
| GenBank mRNA | AK290594.1 | - | 8, 9 | 1039 | ||
| BC028013.1 | - | 8, 9 | 1084 | 5215944 | ||
| M83221.1 | - | 8, 9 | 1145 | |||