Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC20A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257180.1 | NP_001244109.1 | 3 | 1074 | ||
NM_001257181.1 | NP_001244110.1 | 3 | 705 | |||
NM_006749.4 | NP_006740.1 | 3 | 1037 | |||
XM_005273613.3 | XP_005273670.1 | 3 | 812 | |||
XM_005273615.3 | XP_005273672.1 | 3 | 812 | |||
XM_006716391.3 | XP_006716454.1 | 4 | 4292 | |||
XM_017013748.1 | XP_016869237.1 | 3 | 928 | |||
XM_017013750.1 | XP_016869239.1 | 4 | 947 | |||
XM_017013751.1 | XP_016869240.1 | 4 | 821 | |||
XM_017013752.1 | XP_016869241.1 | 4 | 1063 | |||
GenBank mRNA | AK291202.1 | - | 3 | 1074 | ||
AK314774.1 | - | 3 | 706 | |||
BC028600.2 | - | 3 | 749 | 4820601 | ||
L20852.1 | - | 2 | 623 |