Search
Search

SLIT3| Gene Symbol: SLIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271946.1 | NP_001258875.1 | 2 | 635 | ||
| NM_003062.3 | NP_003053.1 | 2 | 635 | |||
| XM_017009779.1 | XP_016865268.1 | 2 | 38 | |||
| GenBank mRNA | AB011538.2 | - | 2 | 635 | ||
| AB017169.1 | - | 2 | 478 | |||
| AK310549.1 | - | 2 | 252 | |||
| AY358884.1 | - | 2 | 297 | |||
| BC146759.1 | - | 2 | 635 | 9007197 | ||