Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SNAP25 SIRNA Assays ›
Gene Symbol:
SNAP25
Gene Name:
synaptosome associated protein 25
Gene Aliases:
CMS18, RIC-4, RIC4, SEC9, SNAP, SNAP-25, SUP, bA416N4.2, dJ1068F16.2
Chromosome Location:
Chr.20: 10218694 - 10307420 on Build GRCh38
UniGene ID:
Hs.167317
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS110032
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SNAP25
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001322902.1
NP_001309831.1
3, 4
380
NM_001322903.1
NP_001309832.1
4, 5
476
NM_001322904.1
NP_001309833.1
4, 5
446
NM_001322905.1
NP_001309834.1
4, 5
409
NM_001322906.1
NP_001309835.1
4, 5
403
NM_001322907.1
NP_001309836.1
5, 6
604
NM_001322908.1
NP_001309837.1
5, 6
527
NM_001322909.1
NP_001309838.1
4, 5
394
NM_001322910.1
NP_001309839.1
4, 5
501
NM_003081.4
NP_003072.2
3, 4
349
NM_130811.3
NP_570824.1
3, 4
349
XM_005260808.4
XP_005260865.1
3, 4
427
XM_017028021.1
XP_016883510.1
4, 5
279
XM_017028022.1
XP_016883511.1
4, 5
371
XM_017028023.1
XP_016883512.1
3, 4
453
GenBank mRNA
AK094560.1
-
3, 4
317
AK098347.1
-
4, 5
222
AK098767.1
-
3, 4
219
AK223617.1
-
3, 4
311
AK289647.1
-
5, 6
510
AK314359.1
-
3, 4
260
BC010647.1
-
3, 4
238
3867544
BI552777.1
-
3, 4
322
BI604120.1
-
3, 4
326
BT019684.1
-
2, 3
106
BX647443.1
-
3, 4
221
D21267.1
-
3, 4
311
DA072367.1
-
3, 4
349
DA162693.1
-
3, 4
453
L19760.1
-
3, 4
194
L19761.1
-
3, 4
194
Back To Top
More Information
Panther Classification:
Molecular Function -
SNARE protein
Gene Ontology Categories:
Function(s)
Process(es)
neurotransmitter uptake
chemical synaptic transmission
neurotransmitter secretion
locomotory behavior
associative learning
regulation of neuron projection development
glutamate secretion
synaptic vesicle exocytosis
synaptic vesicle docking
synaptic vesicle priming
synaptic vesicle fusion to presynaptic active zone membrane
regulation of insulin secretion
long-term synaptic potentiation
SNAP receptor activity
protein binding
syntaxin-1 binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0